6-Oxo-1,4,5,6-tetrahydropyridazine-3-carboxylic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

T2955-1G T2955-5G
TCT2955-1GEA 64.46 CAD
TCT2955-1G TCT2955-5G
6-Oxo-1,4,5,6-tetrahydropyridazine-3-carboxylic acid ≥98.0% (by HPLC, titration analysis)
6-Oxo-1,4,5,6-tetrahydropyridazine-3-carboxylic acid

CAS Number: 27372-38-9
MDL Number: MFCD00052030
Molecular Formula: C5H6N2O3
Molecular Weight: 142.11
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 196
Lambda max.: 257 nm (H2O)

Formula: C₅H₆N₂O₃
Melting Pt: 196 °C
MDL Number: MFCD00052030
CAS Number: 27372-38-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bcb6db4938aff9a02785ef1ad3f5b2f38693843c&vid=bcb6db4938aff9a02785ef1ad3f5b2f38693843c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR