Tetrahydroxydiborane

Supplier: TCI America

Synonyms: Tetrahydroxydiboron, Hypodiboric Acid, Diboronic Acid

T2953-25G T2953-5G
TCT2953-25GEA 706.26 CAD
TCT2953-25G TCT2953-5G
Tetrahydroxydiborane
Tetrahydroxydiborane

CAS Number: 13675-18-8
MDL Number: MFCD05663888
Molecular Formula: H4B2O4
Molecular Weight: 89.65
Form: Crystal
Color: White
Melting point (°C): 290


Other Product Names / Synonyms: Tetrahydroxydiboron, Diboronic Acid, Hypodiboric Acid

Formula: H₄B₂O₄
MW: 89.65 g/mol
Melting Pt: 290 °C
MDL Number: MFCD05663888
CAS Number: 13675-18-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a60b369a3c8185cef1106be61d891084d61f17e3&vid=a60b369a3c8185cef1106be61d891084d61f17e3&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR