Tripyrrolidinophosphine oxide ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Tri(pyrrolidin-1-yl)phosphine oxide

T2952-25G T2952-5G
TCT2952-25GEA 616.52 CAD
TCT2952-25G TCT2952-5G
Tripyrrolidinophosphine oxide ≥98.0% (by GC)
Tripyrrolidinophosphine oxide

CAS Number: 6415-07-2
MDL Number: MFCD00014095
Molecular Formula: C12H24N3OP
Molecular Weight: 257.32
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Very Pale Yellow
Boiling point (°C): 133
Flash Point (°C): 113
Specific Gravity (20/20): 1.12


Other Product Names / Synonyms: Tris(N,N-tetramethylene)phosphoric Triamide

Formula: C₁₂H₂₄N₃OP
MW: 257.32 g/mol
Boiling Pt: 133 °C
Density: 1.12 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00014095
CAS Number: 6415-07-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5e37989b098e3f1fcc01c86c7fa0c87017ea3a6c&vid=5e37989b098e3f1fcc01c86c7fa0c87017ea3a6c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR