3,3',5,5'-Tetramethylbiphenyl ≥98.0%

Supplier: TCI America

Synonyms: 3,3',5,5'-Tetramethyl-1,1'-biphenyl

T2948-1G T2948-5G
TCT2948-1GEA 97.96 CAD
TCT2948-1G TCT2948-5G
3,3',5,5'-Tetramethylbiphenyl ≥98.0%
3,3',5,5'-Tetramethylbiphenyl

CAS Number: 25570-02-9
MDL Number: MFCD00051910
Molecular Formula: C16H18
Molecular Weight: 210.32
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 50

Formula: C₁₆H₁₈
Melting Pt: 50 °C
MDL Number: MFCD00051910
CAS Number: 25570-02-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=81d16ab0c740d19bc666cb617eb78a4ffd2408b6&vid=81d16ab0c740d19bc666cb617eb78a4ffd2408b6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR