1,2,3,9-Tetrahydro-9-methyl-4H-carbazole-4-one ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 1,2,3,4-Tetrahydro-9-methyl-4-oxocarbazole, 1,2,3,4-Tetrahydro-9-methylcarbazol-4-one, Ondansetron Related Compound C

T2942-25G T2942-5G
TCT2942-25GEA 234.79 CAD
TCT2942-25G TCT2942-5G
1,2,3,9-Tetrahydro-9-methyl-4H-carbazole-4-one ≥98.0% (by GC)
1,2,3,9-Tetrahydro-9-methyl-4H-carbazole-4-one

CAS Number: 27387-31-1
MDL Number: MFCD00173748
Molecular Formula: C13H13NO
Molecular Weight: 199.25
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 201


Other Product Names / Synonyms: 1,2,3,4-Tetrahydro-9-methyl-4-oxocarbazole

Formula: C₁₃H₁₃NO
MW: 199.2500 g/mol
Melting Pt: 201 °C
MDL Number: MFCD00173748
CAS Number: 27387-31-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=26205b4865c962514cd6a991215207f3d4d855e8&vid=26205b4865c962514cd6a991215207f3d4d855e8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR