n-Tetratriacontane ≥99.0%

Supplier: TCI America

Synonyms: Tetratriacontane

T2938-1G T2938-5G
TCT2938-1GEA 265.28 CAD
TCT2938-1G TCT2938-5G
n-Tetratriacontane ≥99.0%
n-Tetratriacontane

CAS Number: 14167-59-0
MDL Number: MFCD00009412
Molecular Formula: C34H70
Molecular Weight: 478.93
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 285
Melting point (°C): 74

Formula: H₃C(CH₂)₃₂CH₃
MW: 478.93 g/mol
Boiling Pt: 285 °C
Melting Pt: 74 °C
MDL Number: MFCD00009412
CAS Number: 14167-59-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f25d78a99f05332072afaaf9cb9a5f3f6aeac336&vid=f25d78a99f05332072afaaf9cb9a5f3f6aeac336&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR