2-(2-Thienyl)pyridine ≥98.0%

Supplier: TCI America

Synonyms: 2-(Thiophen-2-yl)pyridine, 2-(2-Pyridyl)thiophene

T2936-25G T2936-5G
TCT2936-25GEA 342.23 CAD
TCT2936-25G TCT2936-5G
2-(2-Thienyl)pyridine ≥98.0%
2-(2-Thienyl)pyridine

CAS Number: 3319-99-1
MDL Number: MFCD00044441
Molecular Formula: C9H7NS
Molecular Weight: 161.22
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: Slightly Pale Yellow
Boiling point (°C): 140
Melting point (°C): 63


Other Product Names / Synonyms: 2-(2-Pyridyl)thiophene

Formula: C₉H₇NS
Boiling Pt: 140 °C
Melting Pt: 63 °C
MDL Number: MFCD00044441
CAS Number: 3319-99-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a4fab058f787eb15ec4e710eb45900e1a53517a5&vid=a4fab058f787eb15ec4e710eb45900e1a53517a5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR