(R)-(-)-1,2,3,4-Tetrahydro-1-naphthylamine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
(R)-1,2,3,4-Tetrahydronaphthalen-1-amine
CAS Number: 23357-46-2
MDL Number: MFCD00671629
Molecular Formula: C10H13N
Molecular Weight: 147.22
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 70
Flash Point (°C): 110
Specific Gravity (20/20): 1.04
Specific rotation [a]20/D: -28 deg (C=2, MeOH)
Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (R)-(-)-1-Amino-1,2,3,4-tetrahydronaphthalene
Formula:
C₁₀H₁₃N MW: 147.22 g/mol Boiling Pt: 70 °C Density: 1.04 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00671629 CAS Number: 23357-46-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...