2-Thiopheneboronic acid pinacol ester ≥98.0%
Supplier: TCI America
Synonyms:
4,4,5,5-Tetramethyl-2-(2-thienyl)-1,3,2-dioxaborolane
CAS Number: 193978-23-3
MDL Number: MFCD05663878
Molecular Formula: C10H15BO2S
Molecular Weight: 210.10
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 68
Lambda max.: 239 nm (MeOH)
Product Categories: Synthetic Organic Chemistry, Boronic Acids & Boronic Acid Derivatives
Other Product Names / Synonyms: 4,4,5,5-Tetramethyl-2-(2-thienyl)-1,3,2-dioxaborolane, 2-Thiopheneboronic Acid Pinacol Ester
Formula:
C₁₀H₁₅BO₂S MW: 210.1 g/mol Melting Pt: 68 °C |
MDL Number:
MFCD05663878 CAS Number: 193978-23-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...