Trimethoxy(1H,1H,2H,2H-nonafluorohexyl)silane ≥97.0% (by GC)

Supplier: TCI America

T2918-25G T2918-5G
TCT2918-25GEA 781.01 CAD
TCT2918-25G TCT2918-5G
Trimethoxy(1H,1H,2H,2H-nonafluorohexyl)silane ≥97.0% (by GC)
Trimethoxy(1H,1H,2H,2H-nonafluorohexyl)silane

CAS Number: 85877-79-8
MDL Number: MFCD08275518
Molecular Formula: C9H13F9O3Si
Molecular Weight: 368.27
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 43
Flash Point (°C): 71
Specific Gravity (20/20): 1.35


Product Categories: Materials Chemistry, Silane Coupling Agents
Other Product Names / Synonyms: Trimethoxy(1H,1H,2H,2H-perfluorohexyl)silane, 1H,1H,2H,2H-Nonafluorohexyltrimethoxysilane

Formula: C₉H₁₃F₉O₃Si
Boiling Pt: 43 °C
Density: 1.35 (20 °C)
Flash Pt: 71 °C
MDL Number: MFCD08275518
CAS Number: 85877-79-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=283c9e6f511f4dbc134bff6954155efb6c1c2f2d&vid=283c9e6f511f4dbc134bff6954155efb6c1c2f2d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR