1,2,3,4-Tetrahydronaphthalene-1-carboxylic acid ≥98.0%
Supplier: TCI America
Synonyms:
1,2,3,4-Tetrahydro-1-naphthoic acid
T2903-25G
T2903-5G
TCT2903-25GEA
344.44
CAD
TCT2903-25G
TCT2903-5G
1,2,3,4-Tetrahydronaphthalene-1-carboxylic acid ≥98.0%
1,2,3,4-Tetrahydronaphthalene-1-carboxylic acid
CAS Number: 1914-65-4
MDL Number: MFCD02179127
Molecular Formula: C11H12O2
Molecular Weight: 176.22
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Boiling point (°C): 135
Melting point (°C): 84
Other Product Names / Synonyms: 1,2,3,4-Tetrahydro-1-naphthoic Acid, Tetralin-1-carboxylic Acid
Formula:
C₁₁H₁₂O₂ Boiling Pt: 135 °C Melting Pt: 84 °C |
MDL Number:
MFCD02179127 CAS Number: 1914-65-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...