N,N,N'-Triphenylbenzidine ≥97.0%

Supplier: TCI America

Synonyms: 4-Diphenylamino-4'-phenylaminobiphenyl

T2891-1G T2891-5G
TCT2891-1GEA 159.58 CAD
TCT2891-1G TCT2891-5G
N,N,N'-Triphenylbenzidine ≥97.0%
N,N,N'-Triphenylbenzidine

CAS Number: 167218-30-6
MDL Number: MFCD17926476
Molecular Formula: C30H24N2
Molecular Weight: 412.54
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Color: Slightly Pale Yellow
Melting point (°C): 167


Product Categories: Materials Chemistry, Building Blocks for Small Molecule Semiconductors, Building Blocks for Small Molecule Semiconductors
Other Product Names / Synonyms: 4-Diphenylamino-4'-phenylaminobiphenyl

Formula: C₃₀H₂₄N₂
MW: 412.54 g/mol
Melting Pt: 167 °C
MDL Number: MFCD17926476
CAS Number: 167218-30-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5591cd9ebf42de3693c55a7cc60e8d048fcc96a3&vid=5591cd9ebf42de3693c55a7cc60e8d048fcc96a3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR