(Trifluoromethyl)tris(triphenylphosphine)copper(I) ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: (Trifluoromethyl)tris(triphenylphosphoranyl)copper, Tris(triphenylphosphine)(trifluoromethyl)copper(I)

T2883-1G T2883-5G
TCT2883-1GEA 586.18 CAD
TCT2883-1G TCT2883-5G
(Trifluoromethyl)tris(triphenylphosphine)copper(I) ≥98.0% (by titrimetric analysis)
(Trifluoromethyl)tris(triphenylphosphine)copper(I)

CAS Number: 325810-07-9
Molecular Formula: C55H45CuF3P3
Molecular Weight: 919.43
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 155


Product Categories: Synthetic Organic Chemistry, Fluorinating Agents, Building Blocks, Fluorinated Bioactive Compounds
Other Product Names / Synonyms: Tris(triphenylphosphine)(trifluoromethyl)copper(I)

Formula: C₅₅H₄₅CuF₃P₃
MW: 922.43 g/mol
Melting Pt: 155 °C
MDL Number: MFCD22200540
CAS Number: 325810-07-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cefbea4a89bb0708c212cd8e258caf9e35de864b&vid=cefbea4a89bb0708c212cd8e258caf9e35de864b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR