(S)-(+)-1,2,3,4-Tetrahydro-1-naphthylamine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: (S)-1,2,3,4-Tetrahydronaphthalen-1-amine

T2878-25G T2878-5G
TCT2878-25GEA 733.12 CAD
TCT2878-25G TCT2878-5G
(S)-(+)-1,2,3,4-Tetrahydro-1-naphthylamine ≥98.0% (by GC, titration analysis)
(S)-(+)-1,2,3,4-Tetrahydro-1-naphthylamine

CAS Number: 23357-52-0
MDL Number: MFCD00671630
Molecular Formula: C10H13N
Molecular Weight: 147.22
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 77
Flash Point (°C): 110
Specific Gravity (20/20): 1.04
Specific rotation [a]20/D: 28 deg (C=2, MeOH)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (S)-(+)-1-Amino-1,2,3,4-tetrahydronaphthalene

Formula: C₁₀H₁₃N
MW: 147.22 g/mol
Boiling Pt: 77 °C
Density: 1.04 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00671630
CAS Number: 23357-52-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=123eb0f57d5139646768a6c8b567c42d078d40f5&vid=123eb0f57d5139646768a6c8b567c42d078d40f5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR