Methyltributylammonium chloride ∼75% in water

Supplier: TCI America

T2873-100ML T2873-500ML
TCT2873-100MLEA 33.18 CAD
TCT2873-100ML TCT2873-500ML
Methyltributylammonium chloride ∼75% in water
Methyltributylammonium chloride

CAS Number: 56375-79-2
MDL Number: MFCD00011847
Molecular Formula: C13H30ClN
Molecular Weight: 235.84
Form: Clear Liquid
Color: Slightly Pale Yellow

Formula: C₁₃H₃₀ClN
MDL Number: MFCD00011847
CAS Number: 56375-79-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7ffb9c62f126d7249c143f16bdf2af185a07e5f2&vid=68aca6fabac62204c6df3d3287a90fa7c0a6491b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR