2,4,6-Tris(2,4,6-tribromophenoxy)-1,3,5-triazine ≥98.0%

Supplier: TCI America

Synonyms: Tris(2,4,6-tribromophenyl) Cyanurate, Cyanuric Acid Tris(2,4,6-tribromophenyl) Ester

T2871-25G T2871-5G
TCT2871-25GEA 256.28 CAD
TCT2871-25G TCT2871-5G
2,4,6-Tris(2,4,6-tribromophenoxy)-1,3,5-triazine ≥98.0%
2,4,6-Tris(2,4,6-tribromophenoxy)-1,3,5-triazine

CAS Number: 25713-60-4
MDL Number: MFCD03092948
Molecular Formula: C21H6Br9N3O3
Molecular Weight: 1067.43
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Color: White
Melting point (°C): 230


Product Categories: Materials Chemistry, Resin Additives
Other Product Names / Synonyms: Cyanuric Acid Tris(2,4,6-tribromophenyl) Ester, Tris(2,4,6-tribromophenyl) Cyanurate

Formula: C₂₁H₆Br₉N₃O₃
MW: 1067.43 g/mol
Melting Pt: 230 °C
MDL Number: MFCD03092948
CAS Number: 25713-60-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de1dc6b7af44d1977abaec7e2e8a21c67f71461&vid=3de1dc6b7af44d1977abaec7e2e8a21c67f71461&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR