N-(Triphenylmethyl)-L-serine methyl ester ≥98.0%
Supplier: TCI America
Synonyms:
(S)-Methyl 3-hydroxy-2-(tritylamino)propanoate, Trt-Ser-OMe, N-Trityl-L-serine Methyl Ester
CAS Number: 4465-44-5
MDL Number: MFCD00192570
Molecular Formula: C23H23NO3
Molecular Weight: 361.44
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 148
Specific rotation [a]20/D: 33 deg (C=1, MeOH)
Product Categories: Bioscience, Amino Acids (N-Protected), Amino Acids (C-Protected)
Other Product Names / Synonyms: N-Trityl-L-serine Methyl Ester, Trt-Ser-OMe
Formula:
C₂₃H₂₃NO₃ MW: 361.44 g/mol Melting Pt: 148 °C |
MDL Number:
MFCD00192570 CAS Number: 4465-44-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...