(3-(Methylamino)propyl)trimethoxysilane ≥95.0%
Supplier: TCI America
Synonyms:
(3-Methylaminopropyl)trimethoxysilane, Trimethoxy[3-(methylamino)propyl]silane, N-Methyl-3-(trimethoxysilyl)propylamine, Trimethoxy(3-(methylamino)propyl)silane
CAS Number: 3069-25-8
MDL Number: MFCD00048021
Molecular Formula: C7H19NO3Si
Molecular Weight: 193.32
Purity/Analysis Method: >95.0% (GC,T)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 106
Flash Point (°C): 82
Specific Gravity (20/20): 0.98
Product Categories: Materials Chemistry, Silane Coupling Agents
Other Product Names / Synonyms: N-Methyl-3-(trimethoxysilyl)propylamine
Formula:
H₃CNHCH₂CH₂CH₂Si(OCH₃)₃ MW: 193.32 g/mol Boiling Pt: 106 °C Density: 0.98 (20 °C) Flash Pt: 82 °C |
MDL Number:
MFCD00048021 CAS Number: 3069-25-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...