1,3,5-Tri(2-thienyl)benzene ≥98.0%
Supplier: TCI America
T2866-1G
T2866-5G
TCT2866-1GEA
170.64
CAD
TCT2866-1G
TCT2866-5G
1,3,5-Tri(2-thienyl)benzene ≥98.0%
1,3,5-Tri(2-thienyl)benzene
CAS Number: 15509-95-2
MDL Number: MFCD02323391
Molecular Formula: C18H12S3
Molecular Weight: 324.47
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: Very Pale Yellow
Boiling point (°C): 298
Melting point (°C): 159
Formula:
C₁₈H₁₂S₃ MW: 324.47 g/mol Boiling Pt: 298 °C Melting Pt: 159 °C |
MDL Number:
MFCD02323391 CAS Number: 15509-95-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...