Triphenylmethylium tetrakis(pentafluorophenyl)borate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

T2863-1G T2863-5G
TCT2863-1GEA 342.39 CAD
TCT2863-1G TCT2863-5G
Triphenylmethylium tetrakis(pentafluorophenyl)borate ≥98.0% (by titrimetric analysis)
Triphenylmethylium tetrakis(pentafluorophenyl)borate

CAS Number: 136040-19-2
MDL Number: MFCD03426981
Molecular Formula: C43H15BF20
Molecular Weight: 922.37
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: Pale Yellow
Melting point (°C): 183


Product Categories: Materials Chemistry, Polymerization Catalysts
Other Product Names / Synonyms: Triphenylcarbenium Tetrakis(pentafluorophenyl)borate, Tritylium Tetrakis(pentafluorophenyl)borate

Formula: C₄₃H₁₅BF₂₀
Melting Pt: 183 °C
MDL Number: MFCD03426981
CAS Number: 136040-19-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fbffde2740c9f8caceed6cbdf51a8fe9414c67ac&vid=fbffde2740c9f8caceed6cbdf51a8fe9414c67ac&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR