Triethoxy(1H,1H,2H,2H-nonafluorohexyl)silane ≥97.0%

Supplier: TCI America

Synonyms: 1H,1H,2H,2H-Nonafluorohexyltriethoxysilane, Triethoxy(1H,1H,2H,2H-perfluorohexyl)silane

T2860-25G T2860-5G
TCT2860-25GEA 216.46 CAD
TCT2860-25G TCT2860-5G
Triethoxy(1H,1H,2H,2H-nonafluorohexyl)silane ≥97.0%
Triethoxy(1H,1H,2H,2H-nonafluorohexyl)silane

CAS Number: 102390-98-7
Molecular Formula: C12H19F9O3Si
Molecular Weight: 410.35
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Colorless
Specific Gravity (20/20): 1.24


Product Categories: Materials Chemistry, Silane Coupling Agents
Other Product Names / Synonyms: 1H,1H,2H,2H-Nonafluorohexyltriethoxysilane, Triethoxy(1H,1H,2H,2H-perfluorohexyl)silane

Formula: C₁₂H₁₉F₉O₃Si
MW: 410.35 g/mol
Density: 1.24 (20 °C)
MDL Number: MFCD09954454
CAS Number: 102390-98-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f14d01ab68d30733cab3a5137bd1157ca21a9039&vid=f14d01ab68d30733cab3a5137bd1157ca21a9039&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR