N,N,N',N'-Tetrakis(2-hydroxyethyl)adipamide ≥98.0% (by GC)
Supplier: TCI America
T2859-25G
T2859-5G
TCT2859-25GEA
197.5
CAD
TCT2859-25G
TCT2859-5G
N,N,N',N'-Tetrakis(2-hydroxyethyl)adipamide ≥98.0% (by GC)
N,N,N',N'-Tetrakis(2-hydroxyethyl)adipamide
CAS Number: 6334-25-4
MDL Number: MFCD07787618
Molecular Formula: C14H28N2O6
Molecular Weight: 320.39
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 128
Other Product Names / Synonyms: N,N,N',N'-Tetrakis(2-hydroxyethyl)hexanediamide
Formula:
C₁₄H₂₈N₂O₆ Melting Pt: 128 °C |
MDL Number:
MFCD07787618 CAS Number: 6334-25-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...