2-Thiophenesulfonamide ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Thiophene-2-sulphonamide, Thiophene-2-sulfonamide

T2851-1G T2851-5G
TCT2851-1GEA 57.83 CAD
TCT2851-1G TCT2851-5G
2-Thiophenesulfonamide ≥98.0% (by GC)
2-Thiophenesulfonamide

CAS Number: 6339-87-3
MDL Number: MFCD00185853
Molecular Formula: C4H5NO2S2
Molecular Weight: 163.21
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 146

Formula: C₄H₅NO₂S₂
MW: 163.21 g/mol
Melting Pt: 146 °C
MDL Number: MFCD00185853
CAS Number: 6339-87-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a6924468f389fec1c896fd62cfb959f7dd8a0bdf&vid=a6924468f389fec1c896fd62cfb959f7dd8a0bdf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR