Trifluoperazine dihydrochloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

T2849-25G T2849-5G
TCT2849-25GEA 549.84 CAD
TCT2849-25G TCT2849-5G
Trifluoperazine dihydrochloride ≥98.0% (by HPLC, titration analysis)
Trifluoperazine dihydrochloride

CAS Number: 440-17-5
MDL Number: MFCD00012656
Molecular Formula: C21H24F3N3S
Molecular Weight: 480.42
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 293
Lambda max.: 312 nm (MeOH)Storage Temperature: <0°C


Product Categories: Synthetic Organic Chemistry, Fluorinated Bioactive Compounds, Bioscience, Neurotransmitter Agent Ingredients
Other Product Names / Synonyms: 10-[3-(4-Methylpiperazin-1-yl)propyl]-2-(trifluoromethyl)phenothiazine Dihydrochloride

Formula: C₂₁H₂₄F₃N₃S
Melting Pt: 293 °C
MDL Number: MFCD00012656
CAS Number: 440-17-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e13ebfed8baa47623efddbe56656da0323a2f9ec&vid=e13ebfed8baa47623efddbe56656da0323a2f9ec&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR