4-(Trifluoromethyl)benzenesulfonamide ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

T2847-1G T2847-5G T2847-25G
TCT2847-1GEA 90.06 CAD
TCT2847-1G TCT2847-5G TCT2847-25G
4-(Trifluoromethyl)benzenesulfonamide ≥98.0% (by HPLC, total nitrogen)
4-(Trifluoromethyl)benzenesulphonamide

CAS Number: 830-43-3
MDL Number: MFCD00159251
Molecular Formula: C7H6F3NO2S
Molecular Weight: 225.19
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Color: White
Melting point (°C): 181


Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives, Fluorinated Building Blocks
Other Product Names / Synonyms: alpha,alpha,alpha-Trifluorotoluene-4-sulfonamide

Formula: C₇H₆F₃NO₂S
Melting Pt: 181 °C
MDL Number: MFCD00159251
CAS Number: 830-43-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=70d3d8f81f202e1fa7abb8179212ee3b88dada1d&vid=70d3d8f81f202e1fa7abb8179212ee3b88dada1d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR