BAY 11-7082 ≥95.0% (by total nitrogen basis)

Supplier: TCI America

T2846-100MG T2846-1G
TCT2846-100MGEA 71.1 CAD
TCT2846-100MG TCT2846-1G
BAY 11-7082 ≥95.0% (by total nitrogen basis)
BAY 11-7082

CAS Number: 19542-67-7
MDL Number: MFCD00712162
Molecular Formula: C10H9NO2S
Molecular Weight: 207.25
Purity/Analysis Method: >95.0% (N)
Form: Crystal
Color: White
Melting point (°C): 134
Storage Temperature: 0-10°C


Product Categories: Bioscience, Immune System, Inflammation, Allergy Research Reagents, Inhibitors (for Pharmacological Research)
Other Product Names / Synonyms: (E)-3-Tosylacrylonitrile, (E)-3-(p-Toluenesulfonyl)acrylonitrile

Formula: C₁₀H₉NO₂S
Melting Pt: 134 °C
MDL Number: MFCD00712162
CAS Number: 19542-67-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e5c64082c27022cad569491e2d5654ffd0ca0261&vid=e5c64082c27022cad569491e2d5654ffd0ca0261&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR