BAPTA tetrasodium salt ≥98.0%

Supplier: TCI America

Synonyms: 1,2-Bis(2-aminophenoxy)ethane-N,N,N',N'-tetraacetic acid tetrasodium Salt, Tetrasodium-1,2-bis(2-aminophenoxy)ethane-N,N,N',N'-tetraacetate, BAPTA-Na4

T2844-1G T2844-200MG
TCT2844-1GEA 376.04 CAD
TCT2844-1G TCT2844-200MG
BAPTA tetrasodium salt ≥98.0%
BAPTA tetrasodium salt

CAS Number: 126824-24-6 MDL Number: MFCD00067691 Molecular Formula: C22H24N2O10 Molecular Weight: 564.37 Purity/Analysis Method: 98.0% (T) Form: Crystal Color: White Storage Temperature: 0°C


Form: Crystal
Color: White
Storage Temperature: <0°C


Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Calcium Chelating Reagents

Formula: C₂₂H₂₄N₂O₁₀
MW: 564.37 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00067691
CAS Number: 126824-24-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a14b3f7aea0f58af8441c62cda2909687f1dbf0&vid=3a14b3f7aea0f58af8441c62cda2909687f1dbf0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR