2-(Trifluoromethyl)pyridine-5-carboxaldehyde ≥98.0%
Supplier: TCI America
Synonyms:
6-(Trifluoromethyl)nicotinaldehyde
T2838-1G
T2838-5G
TCT2838-1GEA
206.66
CAD
TCT2838-1G
TCT2838-5G
2-(Trifluoromethyl)pyridine-5-carboxaldehyde ≥98.0%
2-(Trifluoromethyl)pyridine-5-carboxaldehyde
CAS Number: 386704-12-7
MDL Number: MFCD01862647
Molecular Formula: C7H4F3NO
Molecular Weight: 175.11
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 58
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 6-(Trifluoromethyl)nicotinaldehyde, 5-Formyl-2-(trifluoromethyl)pyridine
Formula:
C7H4F3NO MW: 175.11 g/mol Melting Pt: 58 °C |
MDL Number:
MFCD01862647 CAS Number: 386704-12-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...