Tilorone dihydrochloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 2,7-Bis[2-(diethylamino)ethoxy]-9-fluorenone Dihydrochloride, 2,7-Bis(2-(Diethylamino)ethoxy)-9H-fluoren-9-one dihydrochloride

T2836-100MG T2836-1G
TCT2836-100MGEA 140.62 CAD
TCT2836-100MG TCT2836-1G
Tilorone dihydrochloride ≥98.0% (by HPLC, titration analysis)
Tilorone dihydrochloride

CAS Number: 27591-69-1
MDL Number: MFCD00134071
Molecular Formula: C25H34N2O3
Molecular Weight: 483.47
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: Pale Yellow
Melting point (°C): 234


Product Categories: Bioscience, Antiviral Ingredients
Other Product Names / Synonyms: 2,7-Bis[2-(diethylamino)ethoxy]-9-fluorenone Dihydrochloride

Formula: C₂₅H₃₄N₂O₃
MW: 483.47 g/mol
Melting Pt: 234 °C
MDL Number: MFCD00134071
CAS Number: 27591-69-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9c7476e522da9b2e3656b8842b00a2d40411a0aa&vid=9c7476e522da9b2e3656b8842b00a2d40411a0aa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR