Triclabendazole ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
6-Chloro-5-(2,3-dichlorophenoxy)-2-(methylthio)-1H-benzo[d]imidazole, 5-Chloro-6-(2,3-dichlorophenoxy)-2-(methylthio)benzimidazole, 6-Chloro-5-(2,3-dichlorophenoxy)-2-(methylthio)benzimidazole
CAS Number: 68786-66-3
MDL Number: MFCD00864519
Molecular Formula: C14H9Cl3N2OS
Molecular Weight: 359.65
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 176
Product Categories: Bioscience, Anthelmintic Ingredients
Other Product Names / Synonyms: 5-Chloro-6-(2,3-dichlorophenoxy)-2-(methylthio)benzimidazole, 6-Chloro-5-(2,3-dichlorophenoxy)-2-(methylthio)benzimidazole
Formula:
C₁₄H₉Cl₃N₂OS MW: 359.66 g/mol Melting Pt: 176 °C |
MDL Number:
MFCD00864519 CAS Number: 68786-66-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...