4-Hydroxytetrahydropyran ≥97.0% (by GC)
Supplier: TCI America
Synonyms:
Tetrahydro-2H-pyran-4-ol, Tetrahydro-4H-pyran-4-ol
T2825-1G
T2825-5G
TCT2825-1GEA
36.34
CAD
TCT2825-1G
TCT2825-5G
4-Hydroxytetrahydropyran ≥97.0% (by GC)
4-Hydroxytetrahydropyran
CAS Number: 2081-44-9
MDL Number: MFCD00006633
Molecular Formula: C5H10O2
Molecular Weight: 102.13
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 190
Flash Point (°C): 88
Specific Gravity (20/20): 1.07
Other Product Names / Synonyms: 4-Hydroxytetrahydropyran
Formula:
C₅H₁₀O₂ MW: 102.1317 g/mol Boiling Pt: 190 °C Density: 1.07 (20 °C) Flash Pt: 88 °C |
MDL Number:
MFCD00006633 CAS Number: 2081-44-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...