4-[3-(Trifluoromethyl)-3H-diazirin-3-yl]benzoic acid ≥97.0%
Supplier: TCI America
Synonyms:
4-(1-Azi-2,2,2-trifluoroethyl)benzoic acid, 4-[3-(Trifluoromethyl)-3H-diazirin-3-yl]benzoic Acid
T2820-1G
T2820-200MG
TCT2820-1GEA
989.08
CAD
TCT2820-1G
TCT2820-200MG
4-[3-(Trifluoromethyl)-3H-diazirin-3-yl]benzoic acid ≥97.0%
4-[3-(Trifluoromethyl)-3H-diazirin-3-yl]benzoic acid
CAS Number: 85559-46-2
MDL Number: MFCD00236809
Molecular Formula: C9H5F3N2O2
Molecular Weight: 230.15
Purity/Analysis Method: >97.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 125
Storage Temperature: 0-10°C
Formula:
C₉H₅F₃N₂O₂ MW: 230.15 g/mol Melting Pt: 125 °C |
MDL Number:
MFCD00236809 CAS Number: 85559-46-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...