4-[3-(Trifluoromethyl)-3H-diazirin-3-yl]benzyl alcohol ≥97.0% (by HPLC)

Supplier: TCI America

T2818-1G T2818-200MG
TCT2818-1GEA 464.52 CAD
TCT2818-1G TCT2818-200MG
4-[3-(Trifluoromethyl)-3H-diazirin-3-yl]benzyl alcohol ≥97.0% (by HPLC)
4-[3-(Trifluoromethyl)-3H-diazirin-3-yl]benzyl alcohol

CAS Number: 87736-88-7
Molecular Formula: C9H7F3N2O
Molecular Weight: 216.16
Purity/Analysis Method: >97.0% (HPLC)
Form: Clear Liquid
Color: Pale Yellow
Specific Gravity (20/20): 1.32
Storage Temperature: 0-10°C


Product Categories: Bioscience, Photoaffinity Labels, Crosslinkers
Other Product Names / Synonyms: 3-[4-(Hydroxymethyl)phenyl]-3-(trifluoromethyl)-3H-diazirine

Formula: C₉H₇F₃N₂O
Density: 1.32 (20 °C)
MDL Number: MFCD19442582
CAS Number: 87736-88-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=45bd4b139e156a7dc640cb3ce2c00c891fb761a3&vid=45bd4b139e156a7dc640cb3ce2c00c891fb761a3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR