5,6,7,8-Tetrahydro-1-naphthol ≥98.0% (by GC)
Supplier: TCI America
T2816-25G
T2816-5G
TCT2816-25GEA
323.9
CAD
TCT2816-25G
TCT2816-5G
5,6,7,8-Tetrahydro-1-naphthol ≥98.0% (by GC)
5,6,7,8-Tetrahydro-1-naphthol
CAS Number: 529-35-1
MDL Number: MFCD00001734
Molecular Formula: C10H12O
Molecular Weight: 148.21
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: Very Pale Yellow
Boiling point (°C): 265
Melting point (°C): 70
Formula:
C₁₀H₁₂O MW: 148.2 g/mol Boiling Pt: 265 °C Melting Pt: 70 °C |
MDL Number:
MFCD00001734 CAS Number: 529-35-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...