Nε-trifluoroacetyl-L-lysine ≥98.0%

Supplier: TCI America

Synonyms: H-Lys(Tfa)-OH, ε-TFA-lysine, (S)-2-Amino-6-(2,2,2-trifluoroacetamido)hexanoic acid

T2815-25G T2815-5G
TCT2815-25GEA 90.06 CAD
TCT2815-25G TCT2815-5G
Nε-trifluoroacetyl-L-lysine ≥98.0%
Nepsilon-trifluoroacetyl-L-lysine

CAS Number: 10009-20-8
MDL Number: MFCD00037223
Molecular Formula: C8H13F3N2O3
Molecular Weight: 242.20
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 254
Specific rotation [a]20/D: 19 deg (C=1, 1mol/L HCl)
Storage Temperature: 0-10°C


Product Categories: Bioscience, Side Chain Protected Amino Acids (N,C-termini Free)
Other Product Names / Synonyms: epsilon-TFA-lysine

Formula: C₈H₁₃F₃N₂O₃
MW: 242.20 g/mol
Melting Pt: 254 °C
MDL Number: MFCD00037223
CAS Number: 10009-20-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3e8484c2a5cbc22f5d64d676d9dfc50678139572&vid=3e8484c2a5cbc22f5d64d676d9dfc50678139572&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR