2,4,6-Trifluorophenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

T2804-1G T2804-5G
TCT2804-1GEA 60.04 CAD
TCT2804-1G TCT2804-5G
2,4,6-Trifluorophenylboronic acid (contains varying amounts of Anhydride)
2,4,6-Trifluorophenylboronic acid

CAS Number: 182482-25-3
MDL Number: MFCD01863169
Molecular Formula: C6H4BF3O2
Molecular Weight: 175.90
Form: Crystal
Color: White
Melting point (°C): 235


Other Product Names / Synonyms: 2,4,6-Trifluorobenzeneboronic Acid

Formula: C₆H₄BF₃O₂
Melting Pt: 235 °C
MDL Number: MFCD01863169
CAS Number: 182482-25-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=128a7f0cd4d71a34bb5bd24a54155ab1cdb17c70&vid=128a7f0cd4d71a34bb5bd24a54155ab1cdb17c70&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR