N-(p-Toluenesulfonyl)glycine ≥98.0%

Supplier: TCI America

Synonyms: N-Tosylglycine, N-p-Tosylglycine, Tos-Gly-OH

T2803-25G T2803-5G
TCT2803-25GEA 145.52 CAD
TCT2803-25G TCT2803-5G
N-(p-Toluenesulfonyl)glycine ≥98.0%
N-(p-Toluenesulfonyl)glycine

CAS Number: 1080-44-0
MDL Number: MFCD00045898
Molecular Formula: C9H11NO4S
Molecular Weight: 229.25
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 149


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Tosylglycine, Tos-Gly-OH

Formula: C₉H₁₁NO₄S
Melting Pt: 149 °C
MDL Number: MFCD00045898
CAS Number: 1080-44-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a57895b2f9afb06f6f199a13ab97434750af159&vid=3a57895b2f9afb06f6f199a13ab97434750af159&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR