cis-1,2,3,6-Tetrahydrophthalimide ≥98.0%

Supplier: TCI America

Synonyms: cis-4-Cyclohexene-1,2-dicarboximide, cis-3a,4,7,7a-Tetrahydro-1H-isoindole-1,3(2H)-dione

T2801-250G T2801-25G
TCT2801-250GEA 170.64 CAD
TCT2801-250G TCT2801-25G
cis-1,2,3,6-Tetrahydrophthalimide ≥98.0%
cis-1,2,3,6-Tetrahydrophthalimide

CAS Number: 1469-48-3
MDL Number: MFCD00005880
Molecular Formula: C8H9NO2
Molecular Weight: 151.17
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 138
Flash Point (°C): 178


Other Product Names / Synonyms: cis-4-Cyclohexene-1,2-dicarboximide

Formula: C₈H₉NO₂
Melting Pt: 138 °C
Flash Pt: 178 °C
MDL Number: MFCD00005880
CAS Number: 1469-48-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=968aad4b59eec3df05633a743bb7c879177f5894&vid=968aad4b59eec3df05633a743bb7c879177f5894&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR