2'-Trifluoromethoxyacetophenone ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
1-[2-(Trifluoromethoxy)phenyl]-1-ethanone, 1-(2-(Trifluoromethoxy)phenyl)ethanone
T2794-25G
T2794-5G
TCT2794-25GEA
1624.24
CAD
TCT2794-25G
TCT2794-5G
2'-Trifluoromethoxyacetophenone ≥98.0% (by GC)
2'-Trifluoromethoxyacetophenone
CAS Number: 220227-93-0
MDL Number: MFCD00236320
Molecular Formula: C9H7F3O2
Molecular Weight: 204.15
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Specific Gravity (20/20): 1.29
Formula:
C₉H₇F₃O₂ MW: 204.15 g/mol Density: 1.29 (20 °C) |
MDL Number:
MFCD00236320 CAS Number: 220227-93-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...