2-Hydroxy-4-(trifluoromethyl)pyridine ≥98.0% (by GC)
Supplier: TCI America
T2790-1G
T2790-5G
TCT2790-1GEA
134.3
CAD
TCT2790-1G
TCT2790-5G
2-Hydroxy-4-(trifluoromethyl)pyridine ≥98.0% (by GC)
2-Hydroxy-4-(trifluoromethyl)pyridine
CAS Number: 50650-59-4
Molecular Formula: C6H4F3NO
Molecular Weight: 163.10
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 158
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 4-(Trifluoromethyl)-2-pyridinone, 2-Hydroxy-4-(trifluoromethyl)pyridine
Formula:
C₆H₄F₃NO Melting Pt: 158 °C |
MDL Number:
MFCD00153195 CAS Number: 50650-59-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...