2,2'-[4-(2-Hydroxyethylamino)-3-nitrophenylimino]diethanol ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

T2787-25G T2787-5G
TCT2787-25GEA 193.39 CAD
TCT2787-25G TCT2787-5G
2,2'-[4-(2-Hydroxyethylamino)-3-nitrophenylimino]diethanol ≥98.0% (by HPLC, titration analysis)
2,2'-[4-(2-Hydroxyethylamino)-3-nitrophenylimino]diethanol

CAS Number: 33229-34-4
MDL Number: MFCD00071762
Molecular Formula: C12H19N3O5
Molecular Weight: 285.30
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: Yellow
Melting point (°C): 108


Other Product Names / Synonyms: N1,N4,N4-Tris(2-hydroxyethyl)-2-nitro-1,4-phenylenediamine

Formula: C₁₂H₁₉N₃O₅
Melting Pt: 108 °C
MDL Number: MFCD00071762
CAS Number: 33229-34-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=39757dacbca083577e51317502e37761df7a4610&vid=39757dacbca083577e51317502e37761df7a4610&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR