2,4,6-Triphenyl-1,3,5-triazine ≥98.0% (by GC)

Supplier: TCI America

T2785-1G T2785-5G
TCT2785-1GEA 30.02 CAD
TCT2785-1G TCT2785-5G
2,4,6-Triphenyl-1,3,5-triazine ≥98.0% (by GC)
2,4,6-Triphenyl-1,3,5-triazine

CAS Number: 493-77-6
MDL Number: MFCD00006051
Molecular Formula: C21H15N3
Molecular Weight: 309.37
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 230
Melting point (°C): 236
Lambda max.: 270 nm (CHCl3)

Formula: C₂₁H₁₅N₃
Boiling Pt: 230 °C
Melting Pt: 236 °C
MDL Number: MFCD00006051
CAS Number: 493-77-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9cd6826e9b16eef259a76abec6ea23ab5d0106f7&vid=9cd6826e9b16eef259a76abec6ea23ab5d0106f7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR