6-Nitro-2,3,4-trifluoroaniline ≥98.0% (by GC)

Supplier: TCI America

T2783-1G T2783-5G
TCT2783-1GEA 162.74 CAD
TCT2783-1G TCT2783-5G
6-Nitro-2,3,4-trifluoroaniline ≥98.0% (by GC)
6-Nitro-2,3,4-trifluoroaniline

CAS Number: 148416-38-0
MDL Number: MFCD00174089
Molecular Formula: C6H3F3N2O2
Molecular Weight: 192.10
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: Slightly Pale Yellow
Melting point (°C): 60

Formula: C₆H₃F₃N₂O₂
Melting Pt: 60 °C
MDL Number: MFCD00174089
CAS Number: 148416-38-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e87217369dff500c0cd3ad8ff0e75baf2a3c2865&vid=e87217369dff500c0cd3ad8ff0e75baf2a3c2865&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR