2,3,4,5-Tetrafluoroaniline ≥98.0% (by GC)

Supplier: TCI America

T2782-1G T2782-5G
TCT2782-1GEA 90.85 CAD
TCT2782-1G TCT2782-5G
2,3,4,5-Tetrafluoroaniline ≥98.0% (by GC)
2,3,4,5-Tetrafluoroaniline

CAS Number: 5580-80-3
MDL Number: MFCD00025153
Molecular Formula: C6H3F4N
Molecular Weight: 165.09
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 78
Melting point (°C): 28
Flash Point (°C): 79

Formula: C₆H₃F₄N
MW: 165.09 g/mol
Boiling Pt: 78 °C
Melting Pt: 28 °C
Flash Pt: 79 °C
MDL Number: MFCD00025153
CAS Number: 5580-80-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=129b557eb58e9281ff41732657274ad5ef290c73&vid=129b557eb58e9281ff41732657274ad5ef290c73&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR