5-Thiazolemethanol ≥98.0%

Supplier: TCI America

Synonyms: Thiazol-5-ylmethanol, 5-(Hydroxymethyl)thiazole, Thiazole-5-methanol

T2779-25G T2779-5G
TCT2779-25GEA 312.84 CAD
TCT2779-25G TCT2779-5G
5-Thiazolemethanol ≥98.0%
5-Thiazolemethanol

CAS Number: 38585-74-9
MDL Number: MFCD04115732
Molecular Formula: C4H5NOS
Molecular Weight: 115.15
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Slightly Pale Yellow
Boiling point (°C): 106
Specific Gravity (20/20): 1.32
Storage Temperature: <0°C


Other Product Names / Synonyms: 5-(Hydroxymethyl)thiazole

Formula: C₄H₅NOS
Boiling Pt: 106 °C
Density: 1.32 (20 °C)
MDL Number: MFCD04115732
CAS Number: 38585-74-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d3cad447d2801382f7e569f49a770e5a8b90dc76&vid=d3cad447d2801382f7e569f49a770e5a8b90dc76&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR