3,4,5-Tribromo-1H-pyrazole ≥98.0% (by GC)

Supplier: TCI America

T2777-25G T2777-5G
TCT2777-25GEA 330.22 CAD
TCT2777-25G TCT2777-5G
3,4,5-Tribromo-1H-pyrazole ≥98.0% (by GC)
3,4,5-Tribromo-1H-pyrazole

CAS Number: 17635-44-8
MDL Number: MFCD00040248
Molecular Formula: C3HBr3N2
Molecular Weight: 304.77
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 188

Formula: C₃HBr₃N₂
Melting Pt: 188 °C
MDL Number: MFCD00040248
CAS Number: 17635-44-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a4d369aa550d5474f705e5ca1c563f89d70d001&vid=8a4d369aa550d5474f705e5ca1c563f89d70d001&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR