4-[(Trifluoromethyl)thio]aniline ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 4-((Trifluoromethyl)thio)aniline

T2775-25G T2775-5G
TCT2775-25GEA 403.37 CAD
TCT2775-25G TCT2775-5G
4-[(Trifluoromethyl)thio]aniline ≥98.0% (by GC, titration analysis)
4-[(Trifluoromethyl)thio]aniline

CAS Number: 372-16-7
MDL Number: MFCD00040926
Molecular Formula: C7H6F3NS
Molecular Weight: 193.19
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Color: Very Pale Yellow
Boiling point (°C): 103
Flash Point (°C): 107
Specific Gravity (20/20): 1.35


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 4-Aminophenyl Trifluoromethyl Sulfide

Formula: C₇H₆F₃NS
MW: 193.19 g/mol
Boiling Pt: 103 °C
Density: 1.35 (20 °C)
Flash Pt: 107 °C
MDL Number: MFCD00040926
CAS Number: 372-16-7
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8114b7a8c140822201d12d5b42c83da45e4f95c5&vid=8114b7a8c140822201d12d5b42c83da45e4f95c5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR