5-(Trifluoromethoxy)isatin ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 5-(Trifluoromethoxy)indoline-2,3-dione

T2774-1G T2774-5G
TCT2774-1GEA 77.74 CAD
TCT2774-1G TCT2774-5G
5-(Trifluoromethoxy)isatin ≥98.0% (by GC, titration analysis)
5-(Trifluoromethoxy)isatin

CAS Number: 169037-23-4
MDL Number: MFCD00192524
Molecular Formula: C9H4F3NO3
Molecular Weight: 231.13
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: Pale Yellow
Melting point (°C): 171


Product Categories: Synthetic Organic Chemistry, Isatins (Multi-component Reaction), Fluorinated Building Blocks
Other Product Names / Synonyms: 5-(Trifluoromethoxy)indole-2,3-dione, 5-(Trifluoromethoxy)-2,3-indolinedione

Formula: C₉H₄F₃NO₃
MW: 231.13 g/mol
Melting Pt: 171 °C
MDL Number: MFCD00192524
CAS Number: 169037-23-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=15df7f66574583898e81a5d02983a949508f75bb&vid=15df7f66574583898e81a5d02983a949508f75bb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR