4,5,6,7-Tetrahydrothieno[3,2-c]pyridine hydrochloride ≥98.0%
Supplier: TCI America
T2770-25G
T2770-5G
TCT2770-25GEA
80.58
CAD
TCT2770-25G
TCT2770-5G
4,5,6,7-Tetrahydrothieno[3,2-c]pyridine hydrochloride ≥98.0%
4,5,6,7-Tetrahydrothieno[3,2-c]pyridine hydrochloride
CAS Number: 28783-41-7
MDL Number: MFCD05861485
Molecular Formula: C7H9NS
Molecular Weight: 175.67
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 228
Formula:
C₇H₁₀ClNS Melting Pt: 228 °C |
MDL Number:
MFCD05861485 CAS Number: 28783-41-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...