3-(Trifluoromethyl)-1H-pyrazole ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 3-(Trifluoromethyl)-1H-pyrazole, 3-(Trifluoromethyl)pyrazole

T2765-1G T2765-5G
TCT2765-1GEA 47.4 CAD
TCT2765-1G TCT2765-5G
3-(Trifluoromethyl)-1H-pyrazole ≥98.0% (by GC)
3-(Trifluoromethyl)-1H-pyrazole

CAS Number: 20154-03-4
MDL Number: MFCD00115018
Molecular Formula: C4H3F3N2
Molecular Weight: 136.08
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 98
Melting point (°C): 49

Formula: C₄H₃F₃N₂
MW: 136.08 g/mol
Boiling Pt: 98 °C
Melting Pt: 49 °C
MDL Number: MFCD00115018
CAS Number: 20154-03-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5da0228cc084954393dcc5ad7c80ccb0d6ae1634&vid=5da0228cc084954393dcc5ad7c80ccb0d6ae1634&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR